This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04515626_g1
See other PPP2R5A GE Assays ›
Gene Symbol:
Ppp2r5a
Gene Name:
protein phosphatase 2, regulatory subunit B', alpha
Gene Aliases:
I79_020142
Chromosome Location:
Chr.Un|NW_003615163: 198867 - 222924 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04515626_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp2r5a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_007652131.1 XP_007650321.1 5 - 6 602 60
XM_007652132.1 XP_007650322.1 5 - 6 613 60
XM_007652133.1 XP_007650323.1 6 - 7 698 60
GenBank mRNA JI872285.1 - 4 - 5 401 60
JP052155.1 - 4 - 5 401 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Protein Phosphatase Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0