This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04512810_g1
See other RQCD1 GE Assays ›
Gene Symbol:
Rqcd1
Gene Name:
RCD1 required for cell differentiation1 homolog (S. pombe)
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003614410: 636551 - 654124 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04512810_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rqcd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003507611.2 XP_003507659.1 3 - 4 704 95
GenBank mRNA JI873216.1 - 3 - 4 425 95
JP055892.1 - 3 - 4 425 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0