This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04476793_m1
See other GUCY1A2 GE Assays ›
Gene Symbol:
Gucy1a2
Gene Name:
guanylate cyclase 1, soluble, alpha 2
Gene Aliases:
I79_021923
Chromosome Location:
Chr.Un|NW_003614160: 15145 - 100484 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04476793_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gucy1a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_007646034.1 XP_007644224.1 1 - 2 141 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

guanylate cyclase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header