This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04475173_m1
See other GPR113 GE Assays ›
Gene Symbol:
Gpr113
Gene Name:
G protein-coupled receptor 113
Gene Aliases:
I79_002548
Chromosome Location:
Chr.Un|NW_003613640: 1484725 - 1498573 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04475173_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr113
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003497022.2 XP_003497070.1 1 - 2 543 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0