Assay Details
Gene Symbol: Vamp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505630.2 | XP_003505678.1 | 2 - 3 | 287 | 67 | |
GenBank mRNA | JI888596.1 | - | 2 - 3 | 260 | 67 | |
JP049909.1 | - | 2 - 3 | 260 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
