This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04463638_g1
See other LOC100755848 GE Assays ›
Gene Symbol:
LOC100755848
Gene Name:
fer-1-like protein 4
Gene Aliases:
Fer1l4, I79_007518
Chromosome Location:
Chr.Un|NW_003613833: 1746052 - 1781992 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04463638_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100755848
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003501142.2 XP_003501190.1 2 - 3 134 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0