Assay Details
Gene Symbol: LOC100755848 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501142.2 | XP_003501190.1 | 27 - 28 | 3437 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
