This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04455002_g1
See other LOC100771702 GE Assays ›
Gene Symbol:
LOC100771702
Gene Name:
general transcription factor IIH subunit 2
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003614223: 77963 - 104683 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04455002_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100771702
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003506272.2 XP_003506320.1 10 - 11 827 100
XM_007646760.1 XP_007644950.1 9 - 10 709 100
XM_007646761.1 XP_007644951.1 10 - 11 781 100
XM_007646762.1 XP_007644952.1 10 - 11 859 100
GenBank mRNA JI870764.1 - 10 - 11 825 100
JP037358.1 - 10 - 11 825 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein general transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0