This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04445641_m1
See other AP4S1 GE Assays ›
Gene Symbol:
Ap4s1
Gene Name:
adaptor-related protein complex 4, sigma 1 subunit
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003613718: 231824 - 252207 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04445641_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ap4s1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003499172.2 XP_003499220.1 1 - 2 217 94
XM_007638866.1 XP_007637056.1 1 - 2 217 94
XM_007638873.1 XP_007637063.1 1 - 2 217 94
XM_007638881.1 XP_007637071.1 1 - 2 217 94
XM_007638890.1 XP_007637080.1 1 - 2 217 94
GenBank mRNA JI871025.1 - 1 - 2 96 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

vesicle coat protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0