Assay Details
Gene Symbol: Fdft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497977.2 | XP_003498025.1 | 5 - 6 | 819 | 75 | |
XM_007620414.1 | XP_007618604.1 | 4 - 5 | 871 | 75 | ||
GenBank mRNA | JI890342.1 | - | 5 - 6 | 799 | 75 | |
JP046046.1 | - | 5 - 6 | 798 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
