Assay Details
Gene Symbol: Creld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496857.2 | XP_003496905.1 | 6 - 7 | 1123 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
