This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04422899_m1
See other GPHR GE Assays ›
Gene Symbol:
Gphr
Gene Name:
golgi pH regulator
Gene Aliases:
GPR89, I79_004482
Chromosome Location:
Chr.Un|NW_003613699: 489754 - 528402 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04422899_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gphr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001246813.1 NP_001233742.1 4 - 5 331 103
GenBank mRNA AB362891.1 - 4 - 5 331 103
JI871894.1 - 4 - 5 425 103
JP046668.1 - 4 - 5 425 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Protein Kinase Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0