Assay Details
| Gene Symbol: Gphr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246813.1 | NP_001233742.1 | 12 - 13 | 1101 | 101 | |
| GenBank mRNA | AB362891.1 | - | 12 - 13 | 1101 | 101 | |
| JI871894.1 | - | 12 - 13 | 1195 | 101 | ||
| JP046668.1 | - | 12 - 13 | 1195 | 101 | ||
More Information
Set Membership: |
|

Gphr
Genomic Map