This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04421401_g1
See other TP53 GE Assays ›
Gene Symbol:
Tp53
Gene Name:
tumor protein p53
Gene Aliases:
I79_002739, P53
Chromosome Location:
Chr.Un|NW_003613643: 1638391 - 1643801 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04421401_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tp53
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243976.1 NP_001230905.1 1 - 2 186 67
GenBank mRNA D86070.1 - 1 - 2 186 67
U50395.1 - 1 - 2 172 67
Y08900.1 - 1 - 2 211 67
Y08901.1 - 1 - 2 218 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Apoptosis Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

P53-like transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header