Assay Details
Gene Symbol: Nr4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647110.1 | XP_007645300.1 | 2 - 3 | 1004 | 60 | |
XM_007647111.1 | XP_007645301.1 | 2 - 3 | 826 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
