Assay Details
Gene Symbol: Terf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649312.1 | XP_007647502.1 | 1 - 2 | 114 | 68 | |
GenBank mRNA | JI869334.1 | - | 1 - 2 | 105 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
