Assay Details
Gene Symbol: IL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_538274.4 | XP_538274.1 | 211 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: IL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_538274.4 | XP_538274.1 | 211 | 94 |
Set Membership: |
![]() ![]() ![]() ![]() |