This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02664282_g1
See other BRCC3 GE Assays ›
Gene Symbol:
BRCC3
Gene Name:
BRCA1/BRCA2-containing complex subunit 3
Gene Aliases:
-
Chromosome Location:
Chr.X: 123081262 - 123143509 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02664282_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BRCC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003640241.4 XP_003640289.1 5 - 6 500 89
XM_005642047.3 XP_005642104.1 5 - 6 793 89
XM_005642048.3 XP_005642105.1 4 - 5 670 89
XM_022415679.1 XP_022271387.1 5 - 6 500 89
XM_022415680.1 XP_022271388.1 3 - 4 398 89
XM_022415681.1 XP_022271389.1 5 - 6 500 89
XM_850374.6 XP_855467.3 5 - 6 500 89

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0