Assay Details
| Gene Symbol: MPZL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_546502.6 | XP_546502.3 | 413 | 64 | ||
More Information
Set Membership: |
|
| Gene Symbol: MPZL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_546502.6 | XP_546502.3 | 413 | 64 | ||
Set Membership: |
|