This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02650050_m1
See other NUGGC GE Assays ›
Gene Symbol:
NUGGC
Gene Name:
nuclear GTPase, germinal center associated
Gene Aliases:
C25H8orf80
Chromosome Location:
Chr.25: 29507198 - 29561565 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02650050_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUGGC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022410101.1 XP_022265809.1 12 - 13 2221 70
XM_022410102.1 XP_022265810.1 12 - 13 1691 70
XM_022410103.1 XP_022265811.1 12 - 13 2628 70
XM_022410104.1 XP_022265812.1 12 - 13 2219 70
XM_022410105.1 XP_022265813.1 10 - 11 1477 70
XM_022410106.1 XP_022265814.1 9 - 10 1284 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0