This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02649787_m1
See other CCDC15 GE Assays ›
Gene Symbol:
CCDC15
Gene Name:
coiled-coil domain containing 15
Gene Aliases:
-
Chromosome Location:
Chr.5: 9425436 - 9505370 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02649787_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CCDC15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005619604.3 XP_005619661.1 2140 148
XM_014113321.2 XP_013968796.1 2140 148
XM_014113324.2 XP_013968799.1 2140 148
XM_022418140.1 XP_022273848.1 2253 148
XM_022418141.1 XP_022273849.1 2177 148
XM_022418143.1 XP_022273851.1 2026 148
XM_022418144.1 XP_022273852.1 2017 148
XM_022418145.1 XP_022273853.1 2017 148
XM_022418146.1 XP_022273854.1 1948 148
XM_022418148.1 XP_022273856.1 1979 148

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header