This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02644470_g1
See other APOA2 GE Assays ›
Gene Symbol:
APOA2
Gene Name:
apolipoprotein A2
Gene Aliases:
Apo-AII, ApoA-II
Chromosome Location:
Chr.38: 21263641 - 21265887 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02644470_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APOA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001313799.1 NP_001300728.1 2 - 3 111 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

apolipoprotein

Gene Ontology Categories:

Function(s) Process(es)

phosphatidylcholine biosynthetic process
phospholipid catabolic process
response to glucose
positive regulation of cholesterol esterification
negative regulation of very-low-density lipoprotein particle remodeling
protein oxidation
peptidyl-methionine modification
regulation of intestinal cholesterol absorption
cholesterol efflux
phospholipid efflux
high-density lipoprotein particle clearance
positive regulation of catalytic activity
reverse cholesterol transport
diacylglycerol catabolic process
positive regulation of phagocytosis
protein stabilization
negative regulation of lipid catabolic process
positive regulation of lipid catabolic process
negative regulation of cholesterol import
negative regulation of cholesterol transporter activity
intermembrane lipid transfer
heat shock protein binding
protein homodimerization activity
protein heterodimerization activity
phosphatidylcholine-sterol O-acyltransferase activator activity
intermembrane cholesterol transfer activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0