Assay Details
Gene Symbol: CD3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_536556.5 | XP_536556.1 | 229 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: CD3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_536556.5 | XP_536556.1 | 229 | 74 |
Set Membership: |
![]() ![]() ![]() ![]() |