This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02643635_g1
See other MOCS2 GE Assays ›
Gene Symbol:
MOCS2
Gene Name:
molybdenum cofactor synthesis 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 62153828 - 62164154 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02643635_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOCS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005619329.3 XP_005619386.1 4 - 5 655 99
XM_022417841.1 XP_022273549.1 4 - 5 667 99
XM_022417842.1 XP_022273550.1 4 - 5 632 99
XM_022417843.1 XP_022273551.1 3 - 4 422 99
XM_022417844.1 XP_022273552.1 3 - 4 383 99
XM_022417845.1 XP_022273553.1 4 - 5 671 99
XM_536476.6 XP_536476.1 4 - 5 482 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0