This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02628270_g1
See other IL2RG GE Assays ›
Gene Symbol:
IL2RG
Gene Name:
interleukin 2 receptor subunit gamma
Gene Aliases:
-
Chromosome Location:
Chr.X: 55480846 - 55488485 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02628270_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL2RG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003201.1 NP_001003201.1 4 - 5 592 63
XM_014111314.2 XP_013966789.1 4 - 5 678 63
XM_022415493.1 XP_022271201.1 4 - 5 593 63
GenBank mRNA U04361.1 - 4 - 5 592 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Canis familiaris Inventoried Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

interleukin-15-mediated signaling pathway
positive regulation of phagocytosis
interleukin-15 receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0