This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02622749_g1
See other NPY1R GE Assays ›
Gene Symbol:
NPY1R
Gene Name:
neuropeptide Y receptor Y1
Gene Aliases:
-
Chromosome Location:
Chr.15: 59580546 - 59587753 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02622749_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NPY1R
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002930.1 NP_001002930.1 1 - 2 687 137
XM_005628813.2 XP_005628870.1 2 - 3 2633 137
XM_005628814.2 XP_005628871.1 2 - 3 2039 137
XM_005628817.2 XP_005628874.1 2 - 3 1422 137
XM_005628820.3 XP_005628877.1 2 - 3 932 137
XM_022427650.1 XP_022283358.1 2 - 3 1303 137
GenBank mRNA AF005778.1 - 1 - 2 687 137

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

outflow tract morphogenesis
glucose metabolic process
neuropeptide signaling pathway
locomotory behavior
feeding behavior
regulation of blood pressure
sensory perception of pain
regulation of multicellular organism growth
peptide YY receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0