This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02592255_g1
See other SRA-12 GE Assays ›
Gene Symbol:
sra-12 gly-1
Gene Name:
Serpentine receptor class alpha-12
GLYcosylation related
Gene Aliases:
CELE_F44F4.7
CELE_F44F4.6
Chromosome Location:
Chr.II: 10902756 - 10904479 on Build WBcel235
Chromosome Location:
Chr.II: 10899985 - 10908438 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02592255_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: sra-12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_063945.6 NP_496346.2 5 - 6 876 136
Gene Symbol: gly-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_063944.7 NP_496345.1 5 - 6 1120 136

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_063944.7 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of chemical stimulus
biological_process
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
molecular_function
G-protein coupled receptor activity
olfactory receptor activity
acetylglucosaminyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0