This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02501523_g1
See other TAG-241 GE Assays ›
Gene Symbol:
tag-241
Gene Name:
protein-coding
Gene Aliases:
CELE_C34E11.3
Chromosome Location:
Chr.X: 11824004 - 11836162 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02501523_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tag-241
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001270245.2 NP_001257174.1 23 - 24 4476 116
NM_001368527.5 NP_001355535.1 23 - 24 4340 116
NM_001368528.3 NP_001355536.1 23 - 24 4257 116
NM_001368529.3 NP_001355537.1 22 - 23 4203 116
NM_001375149.2 NP_001361908.1 22 - 23 4214 116
NM_001381110.1 NP_001367232.1 21 - 22 4164 116
NM_001383651.1 23 - 24 4349 116

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0