This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02501512_g1
See other TAG-241 GE Assays ›
Gene Symbol:
tag-241
Gene Name:
protein-coding
Gene Aliases:
CELE_C34E11.3
Chromosome Location:
Chr.X: 11824004 - 11836162 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02501512_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tag-241
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001270245.2 NP_001257174.1 13 - 14 2241 78
NM_001368527.5 NP_001355535.1 13 - 14 2105 78
NM_001368528.3 NP_001355536.1 13 - 14 2097 78
NM_001368529.3 NP_001355537.1 13 - 14 2097 78
NM_001375149.2 NP_001361908.1 13 - 14 2108 78
NM_001381110.1 NP_001367232.1 12 - 13 2058 78
NM_001383651.1 13 - 14 2114 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001375149.2, NM_001383651.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0