Assay Details
Gene Symbol: F52D10.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077536.6 | NP_509937.1 | 2 - 3 | 246 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: F52D10.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077536.6 | NP_509937.1 | 2 - 3 | 246 | 71 |
Set Membership: |
![]() ![]() ![]() ![]() |