Assay Details
Gene Symbol: T25C12.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077518.5 | NP_509919.2 | 26 - 27 | 5581 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: T25C12.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077518.5 | NP_509919.2 | 26 - 27 | 5581 | 78 |
Set Membership: |
![]() ![]() ![]() ![]() |