This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02501069_g1
See other HUM-4 GE Assays ›
Gene Symbol:
hum-4
Gene Name:
protein-coding
Gene Aliases:
CELE_F46C3.3
Chromosome Location:
Chr.X: 11420465 - 11433608 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02501069_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hum-4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001270229.4 NP_001257158.1 18 - 19 3065 83
NM_001270230.4 NP_001257159.1 17 - 18 2798 83
NM_001270232.4 NP_001257161.1 12 - 13 1943 83
NM_001307862.4 NP_001294791.1 17 - 18 2954 83
NM_001307863.4 NP_001294792.1 16 - 17 2687 83
NM_001373506.3 NP_001360186.1 33 6560 83
NM_001373507.3 NP_001359540.1 35 6735 83
NM_001383647.1 33 - 34 6637 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
vesicle transport along actin filament
microfilament motor activity
nucleotide binding
motor activity
actin binding
calmodulin binding
ATP binding
actin-dependent ATPase activity
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0