This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02501011_m1
See other SLO-2 GE Assays ›
Gene Symbol:
slo-2
Gene Name:
protein-coding
Gene Aliases:
CELE_F08B12.3
Chromosome Location:
Chr.X: 11394605 - 11405931 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02501011_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slo-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029356.4 NP_001024527.1 1 - 2 421 96
NM_001029357.5 NP_001024528.1 1 - 2 421 96
GenBank mRNA AF173828.1 - 1 - 2 313 96
AF232770.1 - 1 - 2 114 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of neurotransmitter secretion
potassium ion transport
potassium ion transmembrane transport
intracellular sodium activated potassium channel activity
calcium-activated potassium channel activity
outward rectifier potassium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0