This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02500283_g1
See other PLC-1 GE Assays ›
Gene Symbol:
plc-1
Gene Name:
protein-coding
Gene Aliases:
CELE_F31B12.1
Chromosome Location:
Chr.X: 10794857 - 10838650 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02500283_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: plc-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029446.3 NP_001024617.1 5 - 6 1023 66
NM_001029448.6 NP_001024619.1 5 - 6 985 66
NM_001129669.4 NP_001123141.2 38 - 39 5863 66
NM_001136454.3 NP_001129926.1 5 - 6 985 66
NM_001136455.4 NP_001129927.1 5 - 6 1207 66
NM_001373489.2 NP_001360637.1 38 - 39 5842 66
NM_001373491.2 NP_001360639.1 38 - 39 5872 66
NM_001373492.2 NP_001360635.1 38 - 39 5893 66
NM_001373493.2 NP_001360636.1 39 - 40 6013 66
NM_001373494.2 NP_001359720.1 39 - 40 6034 66
NM_001373495.2 NP_001360638.1 39 - 40 6043 66
NM_077404.6 NP_509805.3 5 - 6 985 66
GenBank mRNA AF044576.1 - 5 - 6 1045 66
AM992255.1 - 5 - 6 985 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

activation of immune response
lipid metabolic process
signal transduction
small GTPase mediated signal transduction
determination of adult lifespan
lipid catabolic process
negative regulation of GTPase activity
intracellular signal transduction
positive regulation of protein import into nucleus
phosphatidylinositol-mediated signaling
defense response to Gram-positive bacterium
regulation of defense response to bacterium, incompatible interaction
1-phosphatidyl-1D-myo-inositol 4,5-bisphosphate catabolic process
phosphatidylinositol phospholipase C activity
guanyl-nucleotide exchange factor activity
GTPase inhibitor activity
phosphoric diester hydrolase activity
hydrolase activity
Ras GTPase binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0