This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02500262_m1
See other PLC-1 GE Assays ›
Gene Symbol:
plc-1
Gene Name:
protein-coding
Gene Aliases:
CELE_F31B12.1
Chromosome Location:
Chr.X: 10794857 - 10838650 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02500262_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: plc-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029446.3 NP_001024617.1 15 - 16 2646 110
NM_001029448.6 NP_001024619.1 15 - 16 2608 110
NM_001129669.4 NP_001123141.2 48 - 49 7486 110
NM_001136454.3 NP_001129926.1 15 - 16 2638 110
NM_001136455.4 NP_001129927.1 15 - 16 2830 110
NM_001373489.2 NP_001360637.1 48 - 49 7465 110
NM_001373491.2 NP_001360639.1 48 - 49 7495 110
NM_001373492.2 NP_001360635.1 48 - 49 7516 110
NM_001373493.2 NP_001360636.1 49 - 50 7636 110
NM_001373494.2 NP_001359720.1 49 - 50 7657 110
NM_001373495.2 NP_001360638.1 49 - 50 7666 110
NM_077404.6 NP_509805.3 15 - 16 2608 110
GenBank mRNA AF044576.1 - 15 - 16 2668 110
AM992255.1 - 15 - 16 2638 110

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

activation of immune response
lipid metabolic process
signal transduction
small GTPase mediated signal transduction
determination of adult lifespan
lipid catabolic process
negative regulation of GTPase activity
intracellular signal transduction
positive regulation of protein import into nucleus
phosphatidylinositol-mediated signaling
defense response to Gram-positive bacterium
regulation of defense response to bacterium, incompatible interaction
1-phosphatidyl-1D-myo-inositol 4,5-bisphosphate catabolic process
phosphatidylinositol phospholipase C activity
guanyl-nucleotide exchange factor activity
GTPase inhibitor activity
phosphoric diester hydrolase activity
hydrolase activity
Ras GTPase binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0