This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02500260_g1
See other PLC-1 GE Assays ›
Gene Symbol:
plc-1
Gene Name:
protein-coding
Gene Aliases:
CELE_F31B12.1
Chromosome Location:
Chr.X: 10794857 - 10838650 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02500260_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: plc-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029446.3 NP_001024617.1 13 - 14 2424 69
NM_001029448.6 NP_001024619.1 13 - 14 2386 69
NM_001129669.4 NP_001123141.2 46 - 47 7264 69
NM_001136454.3 NP_001129926.1 13 - 14 2416 69
NM_001136455.4 NP_001129927.1 13 - 14 2608 69
NM_001373489.2 NP_001360637.1 46 - 47 7243 69
NM_001373491.2 NP_001360639.1 46 - 47 7273 69
NM_001373492.2 NP_001360635.1 46 - 47 7294 69
NM_001373493.2 NP_001360636.1 47 - 48 7414 69
NM_001373494.2 NP_001359720.1 47 - 48 7435 69
NM_001373495.2 NP_001360638.1 47 - 48 7444 69
NM_077404.6 NP_509805.3 13 - 14 2386 69
GenBank mRNA AF044576.1 - 13 - 14 2446 69
AM992255.1 - 13 - 14 2416 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

activation of immune response
lipid metabolic process
signal transduction
small GTPase mediated signal transduction
determination of adult lifespan
lipid catabolic process
negative regulation of GTPase activity
intracellular signal transduction
positive regulation of protein import into nucleus
phosphatidylinositol-mediated signaling
defense response to Gram-positive bacterium
regulation of defense response to bacterium, incompatible interaction
1-phosphatidyl-1D-myo-inositol 4,5-bisphosphate catabolic process
phosphatidylinositol phospholipase C activity
guanyl-nucleotide exchange factor activity
GTPase inhibitor activity
phosphoric diester hydrolase activity
hydrolase activity
Ras GTPase binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0