This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02499961_g1
See other MBOA-7 GE Assays ›
Gene Symbol:
mboa-7
Gene Name:
Lysophospholipid acyltransferase 7
Gene Aliases:
CELE_F14F3.3
Chromosome Location:
Chr.X: 10521792 - 10524565 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02499961_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mboa-7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_077359.7 NP_509760.2 2 - 3 233 93
GenBank mRNA EU016382.1 - 2 - 3 225 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

nematode larval development
lipid metabolic process
phospholipid metabolic process
phosphatidylinositol biosynthetic process
phospholipid biosynthetic process
oviposition
lipid modification
multicellular organism growth
O-acyltransferase activity
transferase activity
transferase activity, transferring acyl groups
lysophospholipid acyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0