This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02499151_g1
See other HIM-4 GE Assays ›
Gene Symbol:
him-4
Gene Name:
protein-coding
Gene Aliases:
CELE_F15G9.4
Chromosome Location:
Chr.X: 9717565 - 9753764 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02499151_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: him-4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029411.3 NP_001024582.1 26 - 27 7594 69
NM_001373449.3 NP_001359724.1 26 7533 69
NM_001373450.2 NP_001360633.1 25 - 26 7239 69
NM_001373451.2 NP_001360634.1 25 - 26 7239 69
NM_077234.7 NP_509635.2 26 - 27 7533 69
GenBank mRNA AF074901.1 - 26 - 27 7533 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
basement membrane disassembly
basement membrane organization
extracellular matrix structural constituent
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0