This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02499025_g1
See other F49E2.5 GE Assays ›
Gene Symbol:
F49E2.5
Gene Name:
protein-coding
Gene Aliases:
CELE_F49E2.5
Chromosome Location:
Chr.X: 9543652 - 9560804 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02499025_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: F49E2.5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029523.5 NP_001024694.1 5 - 6 1044 122
NM_001029524.6 NP_001024695.1 5 - 6 1056 122
NM_001373430.3 NP_001360758.1 5 - 6 1040 122
NM_001373432.4 NP_001360760.1 5 - 6 1037 122
NM_001373433.2 NP_001360761.1 5 - 6 1040 122
NM_001381054.2 NP_001368747.1 5 - 6 995 122
NM_001381055.1 NP_001366703.1 4 - 5 846 122
NM_001381056.1 NP_001366700.1 4 - 5 846 122
NM_001381057.1 NP_001367097.1 4 - 5 846 122
NM_001381070.1 NP_001366701.1 4 - 5 846 122
NM_001383624.1 4 - 5 896 122
NM_001383625.1 5 - 6 1044 122

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0