This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02498016_g1
See other SEC-16A.2 GE Assays ›
Gene Symbol:
sec-16A.2
Gene Name:
protein-coding
Gene Aliases:
CELE_F13B9.1
Chromosome Location:
Chr.X: 8287249 - 8296656 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02498016_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: sec-16A.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029386.5 NP_001024557.1 9 - 10 2883 72
NM_001373387.3 NP_001360731.1 9 - 10 2895 72
NM_001373388.2 NP_001360732.1 8 - 9 2760 72
NM_001373389.3 NP_001360733.1 8 - 9 2760 72
NM_076998.5 NP_509399.2 9 - 10 2883 72
NM_076999.8 NP_509400.3 10 - 11 2958 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s)

endoplasmic reticulum organization
Golgi organization
protein transport
vesicle-mediated transport
COPII vesicle coating
positive regulation of protein exit from endoplasmic reticulum
protein localization to endoplasmic reticulum exit site

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0