This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02497445_g1
See other ADT-2 GE Assays ›
Gene Symbol:
adt-2
Gene Name:
protein-coding
Gene Aliases:
CELE_F08C6.1
Chromosome Location:
Chr.X: 7586209 - 7595416 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02497445_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: adt-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029361.5 NP_001024532.2 8 - 9 1094 92
NM_001375070.2 NP_001361970.1 8 1094 92
GenBank mRNA AB072347.1 - 8 - 9 1094 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
integrin-mediated signaling pathway
determination of adult lifespan
regulation of cell size
extracellular matrix organization
collagen fibril organization
positive regulation of multicellular organism growth
metalloendopeptidase activity
peptidase activity
metallopeptidase activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0