This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02496980_g1
See other C36B7.6 GE Assays ›
Gene Symbol:
C36B7.6 C36B7.5
Gene Name:
Uncharacterized protein
protein-coding
Gene Aliases:
CELE_C36B7.6
CELE_C36B7.5
Chromosome Location:
Chr.X: 7096745 - 7100338 on Build WBcel235
Chromosome Location:
Chr.X: 7089020 - 7098833 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02496980_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C36B7.6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_076795.5 NP_509196.2 9 - 10 1647 67
Gene Symbol: C36B7.5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001383603.1 3 - 4 533 67

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001383603.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s)

IRE1-mediated unfolded protein response

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0