This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02494458_g1
See other HAM-2 GE Assays ›
Gene Symbol:
ham-2
Gene Name:
protein-coding
Gene Aliases:
CELE_C07A12.1
Chromosome Location:
Chr.X: 4554684 - 4559408 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02494458_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ham-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029192.6 NP_001024363.2 3 - 4 608 83
NM_001029193.7 NP_001024364.2 3 - 4 613 83
NM_001307898.3 NP_001294827.1 2 - 3 421 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cell fate determination
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell migration
regulation of neuron differentiation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
nucleic acid binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0