This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02493636_g1
See other RGS-7 GE Assays ›
Gene Symbol:
rgs-7
Gene Name:
protein-coding
Gene Aliases:
CELE_F56B6.2
Chromosome Location:
Chr.X: 3542542 - 3557783 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02493636_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rgs-7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029543.5 NP_001024714.1 5 - 6 1129 88
NM_001373260.4 NP_001360776.1 7 1294 88
NM_001373261.4 NP_001360777.1 6 - 7 1294 88
NM_001373262.3 NP_001360778.1 5 - 6 1205 88
NM_076209.7 NP_508610.3 6 - 7 1294 88
GenBank mRNA AB080201.1 - 5 - 6 1160 88
AY569308.1 - 6 - 7 1205 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of signal transduction
positive regulation of GTPase activity
GTPase activator activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0