Assay Details
| Gene Symbol: meg-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075966.5 | NP_508367.1 | 5 - 6 | 1982 | 60 | |
More Information
Set Membership: |
|

meg-3| Gene Symbol: meg-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075966.5 | NP_508367.1 | 5 - 6 | 1982 | 60 | |
Set Membership: |
|