This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02489917_m1
See other LGC-55 GE Assays ›
Gene Symbol:
lgc-55
Gene Name:
Ligand-Gated ion Channel
Gene Aliases:
CELE_Y113G7A.5
Chromosome Location:
Chr.V: 20023046 - 20036667 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02489917_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: lgc-55
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_075469.6 NP_507870.2 1 - 2 82 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ion transport
chloride transport
signal transduction
chemical synaptic transmission
mechanosensory behavior
ion transmembrane transport
regulation of membrane potential
neurological system process
chloride transmembrane transport
transmembrane signaling receptor activity
ion channel activity
extracellular ligand-gated ion channel activity
chloride channel activity
neurotransmitter receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0