This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02489178_g1
See other Y69H2.3 GE Assays ›
Gene Symbol:
Y69H2.3
Gene Name:
protein-coding
Gene Aliases:
CELE_Y69H2.3
Chromosome Location:
Chr.V: 18656088 - 18664126 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02489178_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Y69H2.3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001129590.4 NP_001123062.1 2 - 3 166 61
NM_001276997.4 NP_001263926.1 2 - 3 166 61
NM_001276998.3 NP_001263927.1 2 - 3 166 61
NM_001276999.4 NP_001263928.1 2 - 3 157 61
NM_001373188.2 NP_001360620.1 2 - 3 166 61
NM_001373189.3 NP_001360619.1 3 174 61
NM_075241.7 NP_507642.2 2 - 3 164 61
NM_075242.6 NP_507643.3 2 - 3 164 61
NM_075243.8 NP_507644.2 2 - 3 166 61
NM_075244.8 NP_507645.2 2 - 3 164 61
NR_023988.1 2 - 3 144 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0