Assay Details
Gene Symbol: ret-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129554.5 | NP_001123026.1 | 6 - 7 | 9234 | 100 | |
NM_001136438.3 | NP_001129910.1 | 1 - 2 | 819 | 100 | ||
NM_001380820.1 | NP_001366847.1 | 4 - 5 | 1312 | 100 | ||
NM_001383456.1 | 5 - 6 | 1773 | 100 | |||
NM_074255.7 | NP_506656.2 | 6 - 7 | 9335 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380820.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
