This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02482932_g1
See other SEL-1 GE Assays ›
Gene Symbol:
sel-1 mrps-5
Gene Name:
Suppressor/Enhancer of Lin-12
Putative 28S ribosomal protein S5, mitochondrial
Gene Aliases:
CELE_F45D3.5
CELE_E02A10.1
Chromosome Location:
Chr.V: 12559772 - 12563462 on Build WBcel235
Chromosome Location:
Chr.V: 12561634 - 12567299 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02482932_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: sel-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_073743.7 NP_506144.1 6 - 7 1757 99
GenBank mRNA U50829.1 - 5 - 6 1659 99
Gene Symbol: mrps-5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_073744.6 NP_506145.2 2 - 3 893 99

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_073744.6 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
ER-associated ubiquitin-dependent protein catabolic process
IRE1-mediated unfolded protein response
negative regulation of Notch signaling pathway
translation
ubiquitin-protein transferase activity
RNA binding
structural constituent of ribosome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0