This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02481913_g1
See other LGC-44 GE Assays ›
Gene Symbol:
lgc-44
Gene Name:
Ligand-Gated ion Channel
Gene Aliases:
CELE_F46F3.2
Chromosome Location:
Chr.V: 11669338 - 11672597 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02481913_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: lgc-44
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_073553.6 NP_505954.2 6 - 7 1284 152

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ion transport
signal transduction
chemical synaptic transmission
ion transmembrane transport
regulation of membrane potential
neurological system process
chloride transmembrane transport
transmembrane signaling receptor activity
ion channel activity
extracellular ligand-gated ion channel activity
chloride channel activity
neurotransmitter receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0